Learn More
New England Biolabs, Inc. LyoPrime Luna Probe One-Step RT-qPCR Mix with UDG - 120 rxns
SDP
Catalog No. 502237902
Encompass
The LyoPrime Luna Probe One-Step RT-qPCR Mix with UDG is offered in a lyophilized format allowing for ambient/room temperature shipping and storage prior to use. The mix contains all necessary components for one-step RT-qPCR except primers probe(s) and RNA sample. It can be reconstituted by simply adding water to a 2-4X concentration allowing for increased sample input. Performance in multiplexing applications has been optimized with sensitive linear detection achieved for up to 5 targets across a range of inputs.
This item is not returnable. View return policy
Catalog No. 50-223-7902 Supplier New England Biolabs, Inc. Supplier No. L4001S
The LyoPrime Luna Probe One-Step RT-qPCR Mix with UDG is offered in a lyophilized format allowing for ambient/room temperature shipping and storage prior to use. The mix contains all necessary components for one-step RT-qPCR except primers probe(s) and RNA sample. It can be reconstituted by simply adding water to a 2-4X concentration allowing for increased sample input. Performance in multiplexing applications has been optimized with sensitive linear detection achieved for up to 5 targets across a range of inputs.