Learn More
Meridian Life Science Lyo-Ready 1-Step RT-qPCR, 100ML
SDP
Catalog No. 5000856342
Encompass_Preferred
Lyo-Ready 1-Step RT-qPCR Buffer is a glycerol-free qPCR mix containing RNase Inhibitor, reaction buffer, dNTP, MgCl2 and lyo-excipients. In order to produce room temperature lyophilized RT-qPCR reagents, Reverse Transcriptase, Taq polymerase, assay specific primers and probes are added to Lyo-Ready 1-Step RT-qPCR Buffer for subsequent lyophilization.
Catalog No. 50-008-56342 Supplier Meridian Life Science Supplier No. MDX052100
Lyo-Ready 1-Step RT-qPCR Buffer is a glycerol-free qPCR mix containing RNase Inhibitor, reaction buffer, dNTP, MgCl2 and lyo-excipients. In order to produce room temperature lyophilized RT-qPCR reagents, Reverse Transcriptase, Taq polymerase, assay specific primers and probes are added to Lyo-Ready 1-Step RT-qPCR Buffer for subsequent lyophilization.