Learn More
Meridian Life Science Lyo-Ready 1-Step RT-qPCR, 100ML
Supplier: Meridian Life Science MDX062100C
Lyo-Ready 1-Step RT-qPCR Virus Reaction Mix can be used for the development of both RNA and DNA based molecular diagnostic tests, it requires the addition of a lyo-compatible thermostable reverse transcriptasex for amplification of RNA viruses with a high secondary structure.