Learn More
Meridian Life Science Lyo-Ready 1-Step RT-qPCR, 10ML
SDP
Supplier:  Meridian Life Science MDX06210C
Encompass_Preferred
Lyo-Ready 1-Step RT-qPCR Virus Reaction Mix can be used for the development of both RNA and DNA based molecular diagnostic tests, it requires the addition of a lyo-compatible thermostable reverse transcriptasex for amplification of RNA viruses with a high secondary structure.
Catalog No. 50-008-56404