Learn More
Meridian Life Science Lyo-Ready 1-Step RT-qPCR, 1ML
SDP
Catalog No. 5000856428
Encompass_Preferred
Lyo-Ready 1-Step RT-qPCR Virus Mix can be used for the development of both RNA and DNA based molecular diagnostic tests, it contains a separate lyo-compatible thermostable reverse transcriptasex that has been optimized for amplification of RNA viruses with a high secondary structure (reverse transcriptase remains active at 55-60 C).
Catalog No. 50-008-56428 Supplier Meridian Life Science Supplier No. MDX0621
Lyo-Ready 1-Step RT-qPCR Virus Mix can be used for the development of both RNA and DNA based molecular diagnostic tests, it contains a separate lyo-compatible thermostable reverse transcriptasex that has been optimized for amplification of RNA viruses with a high secondary structure (reverse transcriptase remains active at 55-60 C).