Learn More
Meridian Life Science Lyo-Ready 1-Step RT-qPCR Mix, 100ML
SDP
Catalog No. 5000856283
Encompass_Preferred
Lyo-Ready 1-Step RT-qPCR Mix, 2x is a glycerol-free formulation that contains Taq polymerase, RNase Inhibitor, reaction buffer, dNTP, MgCl2 and lyo-excipients and separate reverse transcriptase and dilution buffer. It is ideal for developing room temperature, lyophilized, multiplex RT-qPCR tests and is suited for high-throughput, automated platforms requiring milli-second rehydration times. It can be used as a wet mix or it can be lyophilized without impacting its performance post rehydration.
Catalog No. 50-008-56283 Supplier Meridian Life Science Supplier No. MDX024100
Lyo-Ready 1-Step RT-qPCR Mix, 2x is a glycerol-free formulation that contains Taq polymerase, RNase Inhibitor, reaction buffer, dNTP, MgCl2 and lyo-excipients and separate reverse transcriptase and dilution buffer. It is ideal for developing room temperature, lyophilized, multiplex RT-qPCR tests and is suited for high-throughput, automated platforms requiring milli-second rehydration times. It can be used as a wet mix or it can be lyophilized without impacting its performance post rehydration.