Learn More
Meridian Life Science Lyo-Ready 1-Step RT-qPCR ROX, 10ML
SDP
Catalog No. 5000856521
Encompass_Preferred
Lyo-Ready 1-Step RT-qPCR ROX Mix, 4x is a glycerol-free, one tube formulation combining the latest advances in buffer chemistry and PCR enhancers, together with an optimized concentration of reverse transcriptase, antibody-mediated hot-start polymerase, dNTPs and MgCl2. Lyo-Ready 1-Step RT-qPCR ROX Mix has been designed for highly reproducible, accurate RNA and DNA target amplification, delivering excellent results in multiplex assays. In order to produce room temperature lyophilized RT-qPCR reagents, assay specific primers and probes can be added to Lyo-Ready 1-Step RT-qPCR ROX Mix for subsequent lyophilization.
Catalog No. 50-008-56521 Supplier Meridian Life Science Supplier No. MDX17410
Lyo-Ready 1-Step RT-qPCR ROX Mix, 4x is a glycerol-free, one tube formulation combining the latest advances in buffer chemistry and PCR enhancers, together with an optimized concentration of reverse transcriptase, antibody-mediated hot-start polymerase, dNTPs and MgCl2. Lyo-Ready 1-Step RT-qPCR ROX Mix has been designed for highly reproducible, accurate RNA and DNA target amplification, delivering excellent results in multiplex assays. In order to produce room temperature lyophilized RT-qPCR reagents, assay specific primers and probes can be added to Lyo-Ready 1-Step RT-qPCR ROX Mix for subsequent lyophilization.