Learn More
Meridian Life Science Lyo-Ready AOF dUTP DNA qPCR, 50ML
SDP
Supplier:  Meridian Life Science MDX32250
Encompass_Preferred
Lyo-Ready AOF dUTP DNA qPCR Blood is a one tube formulation free from animal-origin ingredients that combines the latest advances in buffer chemistry and PCR enhancers, together with an optimized concentration of recombinant antibody-mediated hot start polymerase, dNTPs (including dUTP) and MgCl2. Lyo-Ready AOF dUTP DNA qPCR Blood has been designed for highly reproducible, accurate DNA target amplification, delivering excellent results in multiplex assays, even in the presence of blood samples. The mix is suitable for high-throughput assays where there is a greater risk of false positives from cross-over contamination.
Catalog No. 50-008-56379