Learn More
Meridian Life Science Lyo-Ready AOF dUTP RNA/DNA, 5ML
SDP
Supplier:  Meridian Life Science MDX3025
Encompass_Preferred
Lyo-Ready AOF dUTP RNA/DNA LAMP combines the latest advances in buffer chemistry and reaction enhancers, together with an optimized concentration of strand-displacing polymerase, dNTPs, reverse transcriptase and proprietary lyo-excipients allowing ambient temperature stabilization through lyophilization. dUTP and has been added, for fast, sensitive and highly reproducible RNA and DNA target amplification. In order to produce lyophilized ambient-temperature stable assays, specific primers need to be added to the LAMP mixes prior to lyophilization and magnesium prior to amplification. The mixes do not contain animal-derived components, aligning with evolving regulatory trends toward more sustainable in vitro systems, reducing reliance on animals while offering cost-effective, room-temperature stability through a lyophilization-ready formulation.
Catalog No. 50-008-56378