Learn More
Meridian Life Science Lyo-Ready Direct DNA LAMP, 5ML
SDP
Supplier:  Meridian Life Science MDX1545
Encompass_Preferred
Lyo-Ready Direct DNA LAMP Urine mix combines even greater tolerance with fast, robust isothermal amplification to enable highly sensitive direct detection using crudely processed urine samples. The mix is uniquely designed to overcome much higher concentrations of inhibitors, and so do not require a DNA purification step to concentrate and purify a patient sample. Very fast reaction kinetics result in faster time to results and superior amplification sensitivity and specificity. In addition, the 4x concentrated master mix is formulated with lyo-excipients which are optimized for freeze-drying to create ambient-temperature stable assays, making it ideal for POC urine testing.
Catalog No. 50-008-56441