Learn More
Meridian Life Science Lyo-Ready Direct DNA qPCR, 1ML
SDP
Supplier:  Meridian Life Science MDX1221
Encompass_Preferred
Lyo-Ready DNA qPCR Blood is a 4x qPCR master mix for the development of DNA blood tests, enabling highly sensitive detection of target DNA from whole blood, plasma and serum samples. The mix has been formulated specifically to overcome the inhibitors found in blood samples – no further optimization is required aside from the addition of primers and probes. Furthermore, these mixes can be used in a wet format or lyophilized to create ambient-temperature stable whole blood tests, plasma tests or serum tests.
Catalog No. 50-008-56298