Learn More
Meridian Life Science Lyo-Ready Direct RNA/DNA, 500ML
SDP
Catalog No. 5000856528
Encompass_Preferred
Lyo-Ready RNA/DNA LAMP Saliva mix combines even greater tolerance with fast, robust isothermal amplification to enable highly sensitive direct detection using crudely processed saliva or sputum samples. The mix is uniquely designed to overcome much higher concentrations of inhibitors, and so do not require a DNA or RNA purification step to concentrate and purify a patient sample. Very fast reaction kinetics result in faster time to results and superior amplification sensitivity and specificity. In addition, the 4x concentrated master mix is formulated with lyo-excipients which are optimized for freeze-drying to create ambient-temperature stable assays, making it ideal for POC saliva testing.
Catalog No. 50-008-56528 Supplier Meridian Life Science Supplier No. MDX135500
Lyo-Ready RNA/DNA LAMP Saliva mix combines even greater tolerance with fast, robust isothermal amplification to enable highly sensitive direct detection using crudely processed saliva or sputum samples. The mix is uniquely designed to overcome much higher concentrations of inhibitors, and so do not require a DNA or RNA purification step to concentrate and purify a patient sample. Very fast reaction kinetics result in faster time to results and superior amplification sensitivity and specificity. In addition, the 4x concentrated master mix is formulated with lyo-excipients which are optimized for freeze-drying to create ambient-temperature stable assays, making it ideal for POC saliva testing.