Learn More
Meridian Life Science Lyo-Ready Direct RNA/DNA, 5ML
SDP
Catalog No. 5000856531
Encompass_Preferred
Lyo-Ready RNA/DNA qPCR Blood is a 4x qPCR master mix for the development of both DNA and RNA blood tests from whole blood, plasma and serum samples. The mix has been formulated specifically to overcome the inhibitors found in blood samples – no further optimization is required aside from the addition of primers and probes. Furthermore, these mixes can be used in a wet format or lyophilized to create ambient-temperature stable whole blood tests, plasma tests or serum tests.
Catalog No. 50-008-56531 Supplier Meridian Life Science Supplier No. MDX1235
Lyo-Ready RNA/DNA qPCR Blood is a 4x qPCR master mix for the development of both DNA and RNA blood tests from whole blood, plasma and serum samples. The mix has been formulated specifically to overcome the inhibitors found in blood samples – no further optimization is required aside from the addition of primers and probes. Furthermore, these mixes can be used in a wet format or lyophilized to create ambient-temperature stable whole blood tests, plasma tests or serum tests.