Learn More
Meridian Life Science Lyo-Ready dU 1-Step RT-qPCR, 100ML
SDP
Catalog No. 5000856239
Encompass_Preferred
Lyo-Ready dUTP 1-Step RT-qPCR Reaction Mix combines the latest advances in buffer chemistry and PCR enhancers, together with hot start polymerase and optimized dNTP/dUTP mix. With the addition of a glycerol-free reverse transcriotase, it is designed for stringent amplification of low copy-number RNA and DNA targets and is ideal for high-throughput, automated platforms. It can be used as a wet mix or it can be lyophilized without impacting its performance post rehydration.
Catalog No. 50-008-56239 Supplier Meridian Life Science Supplier No. MDX113100A
Lyo-Ready dUTP 1-Step RT-qPCR Reaction Mix combines the latest advances in buffer chemistry and PCR enhancers, together with hot start polymerase and optimized dNTP/dUTP mix. With the addition of a glycerol-free reverse transcriotase, it is designed for stringent amplification of low copy-number RNA and DNA targets and is ideal for high-throughput, automated platforms. It can be used as a wet mix or it can be lyophilized without impacting its performance post rehydration.