Learn More
Meridian Life Science Lyo-Ready Genotyping Direct, 1ML
SDP
Catalog No. 5000856364
Encompass_Preferred
Lyo-Ready Genotyping Direct qPCR Stool is a mix that is designed for fast, precise, and reproducible allelic discrimination and cluster separation. Its advanced formulation enables highly sensitive amplification of SNPs and other point mutations from stool samples and overcomes inhibitors such as bile salts that can damage DNA and inhibit PCR reactions. Furthermore, it can be used in a wet format or lyophilized to create ambient-temperature stable assays, making it ideal for point-of-care (POC) devices.
Catalog No. 50-008-56364 Supplier Meridian Life Science Supplier No. MDX1481
Lyo-Ready Genotyping Direct qPCR Stool is a mix that is designed for fast, precise, and reproducible allelic discrimination and cluster separation. Its advanced formulation enables highly sensitive amplification of SNPs and other point mutations from stool samples and overcomes inhibitors such as bile salts that can damage DNA and inhibit PCR reactions. Furthermore, it can be used in a wet format or lyophilized to create ambient-temperature stable assays, making it ideal for point-of-care (POC) devices.