Learn More
Meridian Life Science Lyo-Ready Genotyping Direct, 1ML
SDP
Supplier:  Meridian Life Science MDX1681
Encompass_Preferred
To minimize the workflow to isolate DNA from FFPE tissue, Meridian has created the Lyo-Ready Genotyping Direct qPCR FFPE, this allows digestion of the tissue in a single tube, without the need for purification, greatly reducing the risk of sample damage, loss and contamination whilst simplifying and accelerating the analysis process. Furthermore, it can be used in a liquid format or lyophilized format to create ambient-temperature stable assays, making it ideal for high throughput.
Catalog No. 50-008-56467