Learn More
Meridian Life Science Lyo-Ready Genotyping Direct, 5ML
Supplier: Meridian Life Science MDX1285
Lyo-Ready Genotyping Direct qPCR Blood is a 4x inhibitor-resistant mix designed for highly specific and sensitive amplification for genotyping sequence variants, even in the presence of PCR inhibitors found in blood, serum or plasma, to give exceptional cluster resolution with clear allele discrimination, even with challenging SNP targets. Furthermore, it can be used in a liquid or lyophilized format to create ambient-temperature stable assays, making it ideal for point-of-care (POC) devices.