Learn More
Meridian Life Science Lyo-Ready LAMP Mix 4x, 50ML
SDP
Catalog No. 5000856238
Encompass_Preferred
Lyo-Ready LAMP Mix 4x is a glycerol-free mix for use in loop-mediated isothermal amplification (LAMP). It combines the latest advances in buffer chemistry and reaction enhancers, together with an optimized concentration of strand-displacing Polymerase, dNTP and proprietary lyo-excipients allowing ambient temperature stabilization through lyophilization. Lyo-Ready LAMP Mix has been designed for fast, sensitive and highly reproducible DNA target amplification. In order to produce lyophilized ambient-temperature stable LAMP assays, specific primers need to be added prior to lyophilization.
Catalog No. 50-008-56238 Supplier Meridian Life Science Supplier No. MDX09750
Lyo-Ready LAMP Mix 4x is a glycerol-free mix for use in loop-mediated isothermal amplification (LAMP). It combines the latest advances in buffer chemistry and reaction enhancers, together with an optimized concentration of strand-displacing Polymerase, dNTP and proprietary lyo-excipients allowing ambient temperature stabilization through lyophilization. Lyo-Ready LAMP Mix has been designed for fast, sensitive and highly reproducible DNA target amplification. In order to produce lyophilized ambient-temperature stable LAMP assays, specific primers need to be added prior to lyophilization.