Learn More
Meridian Life Science Lyo-Ready qPCR Buffer 2.5x, 8ML
SDP
Catalog No. 5000856548
Encompass_Preferred
Lyo-Ready qPCR Buffer 2.5x is a glycerol-free qPCR reaction buffer containing lyo-excipients, designed to deliver highly reproducible, accurate qPCR assay results under fast thermal cycling conditions. In order to produce lyophilized, ambient-temperature stable qPCR reagents, dNTP, MgCl2 and glycerol-free Taq DNA polymerase (such as MDX011 or MDX015) must be added prior to lyophilization.
Catalog No. 50-008-56548 Supplier Meridian Life Science Supplier No. MDX0228
Lyo-Ready qPCR Buffer 2.5x is a glycerol-free qPCR reaction buffer containing lyo-excipients, designed to deliver highly reproducible, accurate qPCR assay results under fast thermal cycling conditions. In order to produce lyophilized, ambient-temperature stable qPCR reagents, dNTP, MgCl2 and glycerol-free Taq DNA polymerase (such as MDX011 or MDX015) must be added prior to lyophilization.