Learn More
Meridian Life Science Lyo-Ready qPCR Buffer w/o, 100ML
SDP
Catalog No. 5000856315
Encompass_Preferred
Lyo-Ready qPCR Buffer w/o Excipients, 4x is a glycerol-free qPCR reaction buffer, designed to deliver highly reproducible, accurate qPCR assay results under fast thermal cycling conditions. In order to produce lyophilized, ambient-temperature stable qPCR reagents, dNTP, MgCl2, glycerol-free Taq DNA polymerase (such as MDX011 or MDX015) and excipients must be added prior to lyophilization.
Catalog No. 50-008-56315 Supplier Meridian Life Science Supplier No. MDX061100
Lyo-Ready qPCR Buffer w/o Excipients, 4x is a glycerol-free qPCR reaction buffer, designed to deliver highly reproducible, accurate qPCR assay results under fast thermal cycling conditions. In order to produce lyophilized, ambient-temperature stable qPCR reagents, dNTP, MgCl2, glycerol-free Taq DNA polymerase (such as MDX011 or MDX015) and excipients must be added prior to lyophilization.