Learn More
Meridian Life Science Lyo-Ready qPCR Mix 2.6x, 100ML
SDP
Catalog No. 5000856251
Encompass_Preferred
Lyo-Ready qPCR Mix, 2.6x is a higher concentration, glycerol-free formulation that contains Taq polymerase, reaction buffer, dNTP, MgCl2 and lyo-excipients. It is ideal for developing room temperature, lyophilized, multiplex qPCR diagnostic tests and is suited for use with microfluidic qPCR, where the initial volume needs to be smaller. It can be used as a wet mix or it can be lyophilized without impacting its performance post rehydration.
Catalog No. 50-008-56251 Supplier Meridian Life Science Supplier No. MDX023100
Lyo-Ready qPCR Mix, 2.6x is a higher concentration, glycerol-free formulation that contains Taq polymerase, reaction buffer, dNTP, MgCl2 and lyo-excipients. It is ideal for developing room temperature, lyophilized, multiplex qPCR diagnostic tests and is suited for use with microfluidic qPCR, where the initial volume needs to be smaller. It can be used as a wet mix or it can be lyophilized without impacting its performance post rehydration.