Learn More
Meridian Life Science Lyo-Ready qPCR Mix, 5ML
SDP
Catalog No. 5000856491
Encompass_Preferred
Lyo-Ready qPCR Mix, 2x is a glycerol-free formulation that contains Taq polymerase, reaction buffer, dNTP, MgCl2 and lyo-excipients. It is ideal for developing room temperature, lyophilized, multiplex qPCR diagnostic tests and is suited for high-throughput, automated platforms requiring milli-second rehydration times. It can be used as a wet mix or it can be lyophilized without impacting its performance post rehydration.
Catalog No. 50-008-56491 Supplier Meridian Life Science Supplier No. MDX0215
Lyo-Ready qPCR Mix, 2x is a glycerol-free formulation that contains Taq polymerase, reaction buffer, dNTP, MgCl2 and lyo-excipients. It is ideal for developing room temperature, lyophilized, multiplex qPCR diagnostic tests and is suited for high-throughput, automated platforms requiring milli-second rehydration times. It can be used as a wet mix or it can be lyophilized without impacting its performance post rehydration.