Learn More
Meridian Life Science Lyo-Ready RT-LAMP 1-Step Mix, 1ML
SDP
Catalog No. 5000856347
Encompass_Preferred
Lyo-Ready RT-LAMP 1-Step Mix is a glycerol-free mix for isothermal applications such as Loop-Mediated Isothermal Amplification (LAMP). It contains Reverse Transcriptase, Bst-DNA Polymerase (exo-), reaction buffer, dNTP and provided with lyo-excipients. This is a robust master mix for loop-mediated isothermal amplification (LAMP) reactions, has been designed for amplification of RNA and DNA targets. In order to produce lyophilized ambient temperature stable RT-LAMP and LAMP, assay specific primers are added to Lyo-Ready RT-LAMP 1-Step Mix for subsequent lyophilization.
Catalog No. 50-008-56347 Supplier Meridian Life Science Supplier No. MDX1081
Lyo-Ready RT-LAMP 1-Step Mix is a glycerol-free mix for isothermal applications such as Loop-Mediated Isothermal Amplification (LAMP). It contains Reverse Transcriptase, Bst-DNA Polymerase (exo-), reaction buffer, dNTP and provided with lyo-excipients. This is a robust master mix for loop-mediated isothermal amplification (LAMP) reactions, has been designed for amplification of RNA and DNA targets. In order to produce lyophilized ambient temperature stable RT-LAMP and LAMP, assay specific primers are added to Lyo-Ready RT-LAMP 1-Step Mix for subsequent lyophilization.