Learn More
Meridian Life Science Lyo-Ready T4 Ligase Buffer, 1ML
SDP
Catalog No. 5000856374
Encompass_Preferred
Lyo-Ready T4 Ligase Buffer, 5x is a glycerol-free ligation reaction buffer containing ATP and lyo-excipients. It is designed to be used with Glycerol-Free T4 DNA Ligase (HC) (MDX200) to create a wet mix or a lyophilized, ambient-temperature stable mix for the ligation of cohesive blunt ended DNA.
Catalog No. 50-008-56374 Supplier Meridian Life Science Supplier No. MDX2041
Lyo-Ready T4 Ligase Buffer, 5x is a glycerol-free ligation reaction buffer containing ATP and lyo-excipients. It is designed to be used with Glycerol-Free T4 DNA Ligase (HC) (MDX200) to create a wet mix or a lyophilized, ambient-temperature stable mix for the ligation of cohesive blunt ended DNA.