Learn More
New England Biolabs, Inc. LyoPrime WarmStart™ Fluorescent LAMP/RT-LAMP Mix (with UDG) - 96 rxns
SDP
Catalog No. 502581814
Encompass
The LyoPrime WarmStart Fluorescent LAMP/RT-LAMP Mix (with UDG) is offered in a lyophilized format, allowing for ambient/room temperature shipping and storage prior to use. It is a simple, one-step solution for real-time detection of LAMP/RT-LAMP reactions via fluorescence. LAMP and RT-LAMP are commonly used isothermal amplification techniques that provide rapid detection of a target nucleic acid using LAMP-specific primers and a strand-displacing DNA polymerase. Featuring a blend of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase in an optimized buffer solution, this mix offers improved performance. LAMP Fluorescent Dye at 0.5X concentration is included to enable detection of amplification via real time fluorescence. Antarctic Thermolabile UDG and dUTP reduce the risk of carryover contamination between reactions. It is available in 2 x 50 reaction glass vials or a 96-well plate (1 reaction per well).
Catalog No. 50-258-1814 Supplier New England Biolabs, Inc. Supplier No. L4401P
The LyoPrime WarmStart Fluorescent LAMP/RT-LAMP Mix (with UDG) is offered in a lyophilized format, allowing for ambient/room temperature shipping and storage prior to use. It is a simple, one-step solution for real-time detection of LAMP/RT-LAMP reactions via fluorescence. LAMP and RT-LAMP are commonly used isothermal amplification techniques that provide rapid detection of a target nucleic acid using LAMP-specific primers and a strand-displacing DNA polymerase. Featuring a blend of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase in an optimized buffer solution, this mix offers improved performance. LAMP Fluorescent Dye at 0.5X concentration is included to enable detection of amplification via real time fluorescence. Antarctic Thermolabile UDG and dUTP reduce the risk of carryover contamination between reactions. It is available in 2 x 50 reaction glass vials or a 96-well plate (1 reaction per well).