Learn More
Wl Gore And Associates GORE LYOGUARD Freeze-Drying Trays, 15/pk
Catalog No. NC1679684
Encompass
Single-use, enclosed freeze-drying trays that keep product in the tray during the lyophilization process. Keeping liquid or lyophilized API in stainless steel trays is a challenging and time-consuming problem during processing. GORE LYOGUARD Freeze-Drying Trays were designed in consultation with industry experts to contain product during the freeze-drying process. For nearly 20 years, GORE LYOGUARD Trays have been trusted to help manufacturers minimize the risk of exposure to drug product and spend less time worrying about product ejection and cleaning.
Catalog No. NC1679684 Supplier Wl Gore And Associates Supplier No. LGT2000
Single-use, enclosed freeze-drying trays that keep product in the tray during the lyophilization process. Keeping liquid or lyophilized API in stainless steel trays is a challenging and time-consuming problem during processing. GORE LYOGUARD Freeze-Drying Trays were designed in consultation with industry experts to contain product during the freeze-drying process. For nearly 20 years, GORE LYOGUARD Trays have been trusted to help manufacturers minimize the risk of exposure to drug product and spend less time worrying about product ejection and cleaning.