Learn More
MOLTOX Molecular Toxicology Inc Sprague-Dawley Rat Liver S9, Phenobarbital/ 5 6 Benzoflavone induced, 0.154M KCL. 2 mL/vial, Store in Freezer
SDP
Catalog No. 50948569
Encompass
Post mitochondrial supernatants prepared from Sprague Dawley male rat liver. Prepared using the treatment schedule of Matushima, et. al. 1976. In: de Serres, F.J, et. al., editors. In Vitro Metabolic Activation in Mutagenesis Testing. Amsterdam (NL) Elsevier/North Holland p. 85-88. MOLTOX S9 preparations and cofactor reagents for use in metabolic activation studies are manufactured to rigorous standards of quality and performance and are used by leading industrial, government and academic laboratories worldwide. *Non Returnable*
Catalog No. 50-948-569 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 1105L.2
Post mitochondrial supernatants prepared from Sprague Dawley male rat liver. Prepared using the treatment schedule of Matushima, et. al. 1976. In: de Serres, F.J, et. al., editors. In Vitro Metabolic Activation in Mutagenesis Testing. Amsterdam (NL) Elsevier/North Holland p. 85-88. MOLTOX S9 preparations and cofactor reagents for use in metabolic activation studies are manufactured to rigorous standards of quality and performance and are used by leading industrial, government and academic laboratories worldwide. *Non Returnable*