Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Lyophilized Chemistry Calibrator

Catalog No. 21280087 Shop All Beckman Coulter Products
Change view
Click to view available options
Level:
1
2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
21-280-087 1
21-280-086 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-280-087 Supplier Beckman Coulter Supplier No. DR00701
Add to Cart
Add to Cart

For use with Beckman Coulter analyzers

Contains:

  • 12 - 5 mL calibrators
  • 12 - 6 mL diliuents
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzer
Type Calibrator
Disposable Limited-use
Level 1
Quantity 132 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.