Learn More
Formumax Scientific Inc LYOPHILIZED CLOPHOSOME -A
Supplier:  Formumax Scientific Inc F70101CAL
Encompass_Preferred
Lyophilized Clophosome® -A - Anionic Clodronate Liposomes (5mL)Lyophilized Clophosome® -A is developed by FormuMax to satisfy the demand for clodronate liposomes with long shelf-life. We offer one year shelf-life guarantee from the time of delivery to the date of reconstitution. Just reconstitute by adding sterile pure water and mix (see more details below and the product sheet). Once reconstituted it is used the same way as the standard liquid Clophosome®-A (F70101C-A).
Catalog No. NC1488572