Learn More
Meridian Life Science Lyophilized NGS Library, 3UN
SDP
Catalog No. 5000856274
Encompass_Preferred
A ready-to-use lyophilized next-generation sequencing library preparation kit for SBS protocols that can be shipped and stored at ambient temperature.
Catalog No. 50-008-56274 Supplier Meridian Life Science Supplier No. MDX219B
A ready-to-use lyophilized next-generation sequencing library preparation kit for SBS protocols that can be shipped and stored at ambient temperature.