Learn More
Atmar Glass Company LYOPHILIZER ADAPTER 24/40
Catalog No. NC0675977
Encompass
Straight Lyophilizer Adapter With 24/40 Joint 3/4 Inch Od Heavy Wall Tubing
Catalog No. NC0675977 Supplier Atmar Glass Company Supplier No. A0052821
Straight Lyophilizer Adapter With 24/40 Joint 3/4 Inch Od Heavy Wall Tubing