Learn More
BUCHI Lyovapor™ L-250 Freeze Dryer
GREENER_CHOICESDP
Catalog No. 01670480 Shop All BUCHI Products
Change view
Click to view available options
Style:
Not Software Compatible
Software Compatible
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
01-670-480 Not Software Compatible
01-670-479 Software Compatible
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-670-480 Supplier BUCHI Supplier No. 11L250IN0000
Add to Cart
Add to Cart
Request a Quote
The Lyovapor™ L-250 features groundbreaking EcoStream technology, making it the most eco-friendly laboratory freeze dryer.
  • -85 °C condenser suitable for aqueous and organic-based solvent mixtures
  • 4.5 L condensing capacity for standard laboratory applications
  • EcoStream technology uses natural coolants
  • Global Warming Potential (GWP) of only 4
  • Reduced heat output and noise emissions in the lab
  • Infinite-control technology offers process control via display, software, and mobile app
  • Sample protection mode prevents sample collapse
Specifications
Type Benchtop Freeze Dryer
Capacity (Metric) 4.5 L
No. of Shelves 0
Temperature Range (Metric) Ambient to -105°C
Material Stainless Steel Coil and Collector Chamber
Style Not Software Compatible
Certifications/Compliance Low Environmental Impact Factor
Plug Type All except North America
Voltage 220/240 V