Learn More
Medchemexpress LLC Lyp-1 tfa | 99.52% | 1106.16 | 5 MG
SDP
Catalog No. 5000422982
Encompass_Preferred
LyP-1 TFA is a cyclic 9 amino acids tumor homing peptide that selectively binds to p32 receptors which are overexpressed in various tumor-associated cells. For research use only, not for sale to patients.
  • Shows a remarkable reduction in plaque formation and plaque occupation rates in the LyP-1-treated group (in vivo studies).
  • Higher apoptotic rate in macrophages released from hypoxic plaques observed after LyP-1 treatment compared to control peptide (in vivo studies).
  • Can be labeled with a near-infrared fluorophore (Cy5.5) for optical imaging.
  • Exhibits lymphatics-specific binding.
Catalog No. 50-004-22982 Supplier Medchemexpress LLC Supplier No. HYP2526A5MG
LyP-1 TFA is a cyclic 9 amino acids tumor homing peptide that selectively binds to p32 receptors which are overexpressed in various tumor-associated cells. For research use only, not for sale to patients.
  • Shows a remarkable reduction in plaque formation and plaque occupation rates in the LyP-1-treated group (in vivo studies).
  • Higher apoptotic rate in macrophages released from hypoxic plaques observed after LyP-1 treatment compared to control peptide (in vivo studies).
  • Can be labeled with a near-infrared fluorophore (Cy5.5) for optical imaging.
  • Exhibits lymphatics-specific binding.