Learn More
Medchemexpress LLC LYP-IN-1 | 1404436-51-6 | MFCD34567163 | 99.8% | 501.91 g/mol | C28H20ClNO6 | 5 MG
SDP
Catalog No. 5000264601
Encompass_Preferred
LYP-IN-1 is a potent, selective inhibitor of lymphoid tyrosine phosphatase used in biochemical and cellular studies to probe T-cell and mast cell signaling. The compound contains an alkyne moiety enabling copper-catalyzed azide-alkyne cycloaddition for probe development and labeling in chemical biology applications.
  • Potent and selective LYP inhibition (Ki 110 nM; IC50 0.259 μM).
  • Alkyne handle for copper-catalyzed click chemistry applications.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO up to 2 mg/mL with mild heating and sonication.
  • Stable when stored as powder at -20°C for extended periods.
Catalog No. 50-002-64601 Supplier Medchemexpress LLC Supplier No. HY1089445MG
LYP-IN-1 is a potent, selective inhibitor of lymphoid tyrosine phosphatase used in biochemical and cellular studies to probe T-cell and mast cell signaling. The compound contains an alkyne moiety enabling copper-catalyzed azide-alkyne cycloaddition for probe development and labeling in chemical biology applications.
  • Potent and selective LYP inhibition (Ki 110 nM; IC50 0.259 μM).
  • Alkyne handle for copper-catalyzed click chemistry applications.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO up to 2 mg/mL with mild heating and sonication.
  • Stable when stored as powder at -20°C for extended periods.