Learn More
Aviva Systems Biology Corp LYPD4 Antibody - N-terminal region (ARP58637_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58637_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYPD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LYPD4. Alternate names: MGC42718, SMRApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-62383