Learn More
Aviva Systems Biology Corp LYPLA1 Antibody - middle region (ARP52148_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYPLA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LYPLA1. Alternate names: APT-1, LPL1, LYSOPLA, hAPT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60688