Learn More
Sino Biological LYPLA2 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 12360MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LYPLA2 (rh LYPLA2; Catalog#12360-H07E; O95372; Met 1-Val 231). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7483