Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ Lyra Direct Strep Molecular Assay
SDP

Catalog No. 23044007 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
96 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-007 96 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-007 Supplier Quidel™ Supplier No. M112
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lyra direct strep assay is RT-PCR IVD for rapid, qualitative detection/differentiation of Streptococcus pyogenes, Streptococcus dysgalactiae, Streptococcus equi, Streptococcus canis nucleic acids isolated from throat swab specimens

  • RT-PCR
  • One-step reagent setup
  • Results in <70 minutes after extraction
  • No freezer necessary
  • Refrigration storage
  • Long shelf-life
  • Flexible testing format
  • Includes 12 x 8 master mix, rehydration solution and liquid process controls

Specifications

Certifications/Compliance FDA approved*
Content And Storage 2°C to 8°C
Control Sets Included
Description Qualitative detection and differentiation of Group A β-hemolytic Streptococcus (Streptococcus pyogenes) and pyogenic Group C and G β-hemolytic Streptococcus nucleic acids isolated from throat swab specimens obtained from patients with signs and symptoms of pharyngitis, such as a sore throat
CE Marker Yes
Clia Complexity High
Detectable Analytes beta-haemolytic Streptococcus (Group A), beta-haemolytic Streptococcus Nucleic Acids
Sample Type Throat swab specimens obtained from patients symptomatic of acute pharyngitis
Sensitivity Group A β-hemolytic Streptococcus 96%, Pyogenic Group C and G β-hemolytic Streptococcus 95%
Type Molecular Testing
For Use With (Application) RT-PCR
For Use With (Equipment) FDA approved for use on Applied Biosystems™ 7500 series of instruments
Includes Tests and controls
Packaging Type Kit
Quantity 96 Tests/Kit
Specificity Group A β-hemolytic Streptococcus 98%, Pyogenic Group C and G β-hemolytic Streptococcus 1.8 x 104 CFU/mL
Test Time <70 min. after extraction
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.