Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ Lyra SARS-CoV-2 Molecular Assay
SDP

Catalog No. 23044027 Shop All QuidelOrtho Products
Change view
Click to view available options
Format:
Control
Microfuge
Microwell
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-044-027 Microwell
23-044-011 Control
23-044-026 Microfuge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-044-027 Supplier Quidel™ Supplier No. M939
Add to Cart
Add to Cart
This item is not returnable. View return policy

For in vitro qualitative detection of human coronavirus SARS-CoV-2 from viral RNA extracted from nasal, nasopharyngeal (NP) or oropharyngeal (OP) swab specimens from patients with signs and symptoms of COVID-19, Assay targets non-structural Polyprotein (pp1ab) of SARS-CoV-2 virus

  • RT-PCR
  • One-step reagent setup
  • Results in <75 minutes after extraction
  • No freezer necessary
  • Refrigration storage
  • Flexible testing format
  • Includes 12 x 8 master mix, rehydration solution and liquid process controls

Specifications

Certifications/Compliance Emergency Use Authorization (EUA)
Content And Storage 2°C to 8°C
Control Sets Included
CE Marker Yes
Clia Complexity High
Detectable Analytes SARS-CoV-2
DoA Calibrators No
Format Microwell
Sensitivity PPA – 92/92 = 100% (95% CI: 95.99 to 100%)
Type Molecular Testing
For Use With (Application) RT-PCR
For Use With (Equipment) Emergency Use Authorization (EUA) for use on bioMerieux NucliSENS™ easyMAG™ system or EMAG system, followed by RT-PCR on one of six thermocyclers: Applied Biosystems™ 7500 Fast Dx, Applied Biosystems 7500 Standard, QuantStudioTM Real-Time PCR Instrument, Roche LightCycler™ 480, BioRad CFX96 Touch, or Qiagen Rotor-Gene Q
Includes Tests and controls
Packaging Type Kit
Quantity 96 Tests/Kit
Specificity NPA – 92/92 = 100% (95% CI: 95.99 to 100%)
Test Time <75 min. after extraction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.