Learn More
Medchemexpress LLC Lys-Ala-pNA | 115846-24-7 | 99.8% | 337.37 | 5 MG
SDP
Catalog No. 5000396673
Encompass_Preferred
Lys-Ala-pNA is a hydrolytic substrate for dipeptidyl peptidase (DPP) II with a Km of 0.42 mM. This product is intended for research use only.
  • Hydrolytic substrate for dipeptidyl peptidase (DPP) II
  • Km of 0.42 mM
  • For research use only
  • Appears as a solid
  • Color is light yellow to yellow
  • Stable under recommended storage conditions
Catalog No. 50-003-96673 Supplier Medchemexpress LLC Supplier No. HYP44685MG
Lys-Ala-pNA is a hydrolytic substrate for dipeptidyl peptidase (DPP) II with a Km of 0.42 mM. This product is intended for research use only.
  • Hydrolytic substrate for dipeptidyl peptidase (DPP) II
  • Km of 0.42 mM
  • For research use only
  • Appears as a solid
  • Color is light yellow to yellow
  • Stable under recommended storage conditions