Learn More
Medchemexpress LLC Lys-Arg-Thr-Leu-Arg-Arg | 121284-21-7 | 829.01 | 25 MG
SDP
Catalog No. 5000397312
Encompass_Preferred
Lys-Arg-Thr-Leu-Arg-Arg (KRTLRR) is a hexapeptide that acts as a substrate of protein kinase C from the EGF receptor. It can be used to determine the activity of protein kinase C.
  • Hexapeptide
  • Substrate of protein kinase C from EGF receptor
  • Used to determine the activity of protein kinase C
  • Increases activity of membrane-bound Ca2+-inhibited protein kinase (in vitro)
  • Induces autophosphorylation (in vitro)
Catalog No. 50-003-97312 Supplier Medchemexpress LLC Supplier No. 25MGHYP2673A
Lys-Arg-Thr-Leu-Arg-Arg (KRTLRR) is a hexapeptide that acts as a substrate of protein kinase C from the EGF receptor. It can be used to determine the activity of protein kinase C.
  • Hexapeptide
  • Substrate of protein kinase C from EGF receptor
  • Used to determine the activity of protein kinase C
  • Increases activity of membrane-bound Ca2+-inhibited protein kinase (in vitro)
  • Induces autophosphorylation (in vitro)