Learn More
Medchemexpress LLC Lys-Arg-Thr-Leu-Arg-Arg acetate | 829.01 | 50 MG
SDP
Catalog No. 5000400282
Encompass_Preferred
Lys-Arg-Thr-Leu-Arg-Arg acetate (KRTLRR acetate) is a hexapeptide that functions as a substrate of EGF receptor protein kinase C. This chemical can be utilized to measure protein kinase C activity, making it a valuable tool in cancer research, particularly in cancer targeted therapy and metabolism studies. It appears as a solid with a molecular formula of C34H68N16O8.xC2H4O2 and a molecular weight of 829.01 (free base). It exhibits high solubility in H2O, at concentrations of 100 mg/mL or greater.
  • Substrate for EGF receptor protein kinase C
  • Used to measure protein kinase C activity
  • Hexapeptide composition
  • Solid appearance
  • Highly soluble in water
Catalog No. 50-004-00282 Supplier Medchemexpress LLC Supplier No. HYP2673A50MG
Lys-Arg-Thr-Leu-Arg-Arg acetate (KRTLRR acetate) is a hexapeptide that functions as a substrate of EGF receptor protein kinase C. This chemical can be utilized to measure protein kinase C activity, making it a valuable tool in cancer research, particularly in cancer targeted therapy and metabolism studies. It appears as a solid with a molecular formula of C34H68N16O8.xC2H4O2 and a molecular weight of 829.01 (free base). It exhibits high solubility in H2O, at concentrations of 100 mg/mL or greater.
  • Substrate for EGF receptor protein kinase C
  • Used to measure protein kinase C activity
  • Hexapeptide composition
  • Solid appearance
  • Highly soluble in water