Learn More
Bachem Lys-Bradykinin Acetate
SDP
Supplier:  Bachem 4003280.0100
Encompass_Preferred
Lys-Bradykinin Acetate, H-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH, 342-10-9, C56H85N17O12, Mr 1188.40, 100mg. Kallidin, [Lys0] Bradykinin. Kallidin is the precursor of the vasodilator bradykinin.
Catalog No. 50-259-576