Learn More
Medchemexpress LLC Lys-bradykinin TFA | 00-00-0 | 99.9% | 1188.38 g/mol (free base) | C56H85N17O12 · xC2HF3O2 | 25 MG
SDP
Catalog No. 5000247787
Encompass_Preferred
Lys-bradykinin TFA is the trifluoroacetate salt form of the decapeptide kallidin (Lys-bradykinin). It is used as a research reagent and acts as a ligand for kallidin and bradykinin receptors, supporting studies in vascular regulation, inflammation, and pain signaling.
  • TFA salt form of lys-bradykinin (kallidin).
  • High purity (≈99.85%).
  • Molecular weight 1188.38 g/mol (free base).
  • Provided in small quantities suitable for biochemical assays; 25 mg size available.
  • Intended for receptor binding and vascular biology research applications.
Catalog No. 50-002-47787 Supplier Medchemexpress LLC Supplier No. HY103293A25MG
Lys-bradykinin TFA is the trifluoroacetate salt form of the decapeptide kallidin (Lys-bradykinin). It is used as a research reagent and acts as a ligand for kallidin and bradykinin receptors, supporting studies in vascular regulation, inflammation, and pain signaling.
  • TFA salt form of lys-bradykinin (kallidin).
  • High purity (≈99.85%).
  • Molecular weight 1188.38 g/mol (free base).
  • Provided in small quantities suitable for biochemical assays; 25 mg size available.
  • Intended for receptor binding and vascular biology research applications.