Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lys-bradykinin TFA | 99.9% | 1188.38 g/mol | C56H85N17O12 · x C2HF3O2 | 50 MG
SDP

Catalog No. 5000247317
Encompass_Preferred

Lys-Bradykinin TFA is the trifluoroacetate salt of Lys-Bradykinin (kallidin), a biologically active nonapeptide ligand for bradykinin/kallidin receptors. Supplied as a solid peptide powder for research use, it is commonly used in studies of vascular regulation, inflammation, and receptor pharmacology.

  • Trifluoroacetate salt of Lys-Bradykinin (kallidin).
  • Purity 99.85%.
  • Molecular weight 1188.38 g/mol.
  • Available as solid powder, including 50 MG size.
  • Storage: powder -80°C for 2 years, -20°C for 1 year; in solvent -80°C for 6 months, -20°C for 1 month.
  • Documentation includes COA, RP-HPLC, LCMS, elemental analysis, and SDS.

Catalog No. 50-002-47317 Supplier Medchemexpress LLC Supplier No. HY103293A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lys-Bradykinin TFA is the trifluoroacetate salt of Lys-Bradykinin (kallidin), a biologically active nonapeptide ligand for bradykinin/kallidin receptors. Supplied as a solid peptide powder for research use, it is commonly used in studies of vascular regulation, inflammation, and receptor pharmacology.

  • Trifluoroacetate salt of Lys-Bradykinin (kallidin).
  • Purity 99.85%.
  • Molecular weight 1188.38 g/mol.
  • Available as solid powder, including 50 MG size.
  • Storage: powder -80°C for 2 years, -20°C for 1 year; in solvent -80°C for 6 months, -20°C for 1 month.
  • Documentation includes COA, RP-HPLC, LCMS, elemental analysis, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.