Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lys-bradykinin trifluoroacetate | 00-00-0 | 99.9% | 1188.38 g/mol | C56H85N17O12·xC2HF3O2 | 100 MG
SDP

Catalog No. 5000244945
Encompass_Preferred

Lys-Bradykinin TFA is the trifluoroacetate salt of the decapeptide kallidin (Lys-bradykinin), supplied as a purified peptide powder for laboratory research. It acts as a ligand for kallidin and bradykinin receptors and is involved in vascular regulation, inflammation, and pain signaling.

  • TFA salt of Lys-bradykinin (kallidin).
  • Decapeptide sequence: Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg.
  • High purity (99.9%).
  • Molecular weight 1188.38 g/mol (free base).
  • Storage: powder - sealed, away from moisture and light; powder storage: -80°C for 2 years, -20°C for 1 year.

Catalog No. 50-002-44945 Supplier Medchemexpress LLC Supplier No. HY103293A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lys-Bradykinin TFA is the trifluoroacetate salt of the decapeptide kallidin (Lys-bradykinin), supplied as a purified peptide powder for laboratory research. It acts as a ligand for kallidin and bradykinin receptors and is involved in vascular regulation, inflammation, and pain signaling.

  • TFA salt of Lys-bradykinin (kallidin).
  • Decapeptide sequence: Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg.
  • High purity (99.9%).
  • Molecular weight 1188.38 g/mol (free base).
  • Storage: powder - sealed, away from moisture and light; powder storage: -80°C for 2 years, -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.