Learn More
Medchemexpress LLC Lys(CO-C3-p-I-Ph)-OMe | 2088426-96-2 | 97.2% | 432.30 g/mol | C17H25IN2O3 | 10 MG
SDP
Catalog No. 5000288358
Encompass_Preferred
Lys(CO-C3-p-I-Ph)-OMe is a pharmacokinetic modifier used in research to improve the pharmacokinetic properties of PSMA-targeting ligands, including increased plasma residence time and altered tissue uptake. It is supplied in purified powder and solution formats suitable for preclinical studies.
  • Improves plasma residence time of PSMA ligands.
  • Increases albumin binding to extend circulation half-life.
  • Reduces off-target salivary gland uptake of modified ligands.
  • Available as powder in multiple mg sizes and as 10 mM solution in DMSO.
  • High purity suitable for research applications.
Catalog No. 50-002-88358 Supplier Medchemexpress LLC Supplier No. HY15811810MG
Lys(CO-C3-p-I-Ph)-OMe is a pharmacokinetic modifier used in research to improve the pharmacokinetic properties of PSMA-targeting ligands, including increased plasma residence time and altered tissue uptake. It is supplied in purified powder and solution formats suitable for preclinical studies.
  • Improves plasma residence time of PSMA ligands.
  • Increases albumin binding to extend circulation half-life.
  • Reduces off-target salivary gland uptake of modified ligands.
  • Available as powder in multiple mg sizes and as 10 mM solution in DMSO.
  • High purity suitable for research applications.