Learn More
Medchemexpress LLC Lys-CoA TFA | 98.5% | 50 MG
SDP
Catalog No. 5000340679
Encompass_Preferred
Lys-CoA TFA is a selective p300 histone acetyltransferase (HAT) inhibitor with an IC50 of 50-500 nM. It demonstrates over 100-fold selectivity for p300 compared to PCAF (IC50=200 μM). Lys-CoA TFA effectively inhibits p300 HAT activity-dependent transcriptional activation.
  • Selective p300 histone acetyltransferase (HAT) inhibitor
  • Displays >100-fold selectivity for p300 over PCAF
  • Inhibits p300 HAT activity-dependent transcriptional activation
  • Induces a senescent phenotype in human normal melanocytes at concentrations of 0.1 and 1 mM over 60 hours
  • Almost completely abolished proliferation, induced high levels of SA-β-Gal, and inhibited cyclin E expression in a dose-dependent manner in normal human melanocytes
  • For research use only
Catalog No. 50-003-40679 Supplier Medchemexpress LLC Supplier No. HY13103550MG
Lys-CoA TFA is a selective p300 histone acetyltransferase (HAT) inhibitor with an IC50 of 50-500 nM. It demonstrates over 100-fold selectivity for p300 compared to PCAF (IC50=200 μM). Lys-CoA TFA effectively inhibits p300 HAT activity-dependent transcriptional activation.
  • Selective p300 histone acetyltransferase (HAT) inhibitor
  • Displays >100-fold selectivity for p300 over PCAF
  • Inhibits p300 HAT activity-dependent transcriptional activation
  • Induces a senescent phenotype in human normal melanocytes at concentrations of 0.1 and 1 mM over 60 hours
  • Almost completely abolished proliferation, induced high levels of SA-β-Gal, and inhibited cyclin E expression in a dose-dependent manner in normal human melanocytes
  • For research use only