Learn More
Medchemexpress LLC Lys-CoA TFA (Lys-coenzyme A) | 98.5% | 1 MG
SDP
Catalog No. 5000341008
Encompass_Preferred
Lys-CoA TFA is a selective p300 histone acetyltransferase (HAT) inhibitor with an IC50 of 50-500 nM. It demonstrates over 100-fold selectivity for p300 compared to PCAF (IC50=200 μM). This compound works by inhibiting p300 HAT activity-dependent transcriptional activation.
  • Selective p300 histone acetyltransferase (HAT) inhibitor
  • IC50 of 50-500 nM for p300
  • Over 100-fold selectivity for p300 compared to PCAF
  • Induces a senescent phenotype in human normal melanocytes at 0.1 and 1 mM concentrations
  • Inhibits cyclin E expression in a dose-dependent manner
Catalog No. 50-003-41008 Supplier Medchemexpress LLC Supplier No. HY1310351MG
Lys-CoA TFA is a selective p300 histone acetyltransferase (HAT) inhibitor with an IC50 of 50-500 nM. It demonstrates over 100-fold selectivity for p300 compared to PCAF (IC50=200 μM). This compound works by inhibiting p300 HAT activity-dependent transcriptional activation.
  • Selective p300 histone acetyltransferase (HAT) inhibitor
  • IC50 of 50-500 nM for p300
  • Over 100-fold selectivity for p300 compared to PCAF
  • Induces a senescent phenotype in human normal melanocytes at 0.1 and 1 mM concentrations
  • Inhibits cyclin E expression in a dose-dependent manner