Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lys-coenzyme A trifluoroacetate | 98.5% | 1108.82 g·mol⁻1 | C33H54F3N10O21P3S | 5 MG
SDP

Catalog No. 5000262796
Encompass_Preferred

Lys-CoA TFA is a bisubstrate-mimic inhibitor of the p300 histone acetyltransferase (HAT). It inhibits p300-dependent acetylation (reported IC50 = 50-500 nM) with >100-fold selectivity over PCAF and is intended for use in biochemical and cell-based studies probing p300-mediated acetylation.

  • Selective p300 histone acetyltransferase (HAT) inhibitor (IC50 = 50-500 nM)
  • Greater than 100-fold selectivity versus PCAF
  • High purity (98.5%) suitable for research applications
  • Molecular formula C33H54F3N10O21P3S
  • Molecular weight 1108.82 g·mol⁻1
  • Supplied as a small-quantity solid for biochemical assays and cell studies

Catalog No. 50-002-62796 Supplier Medchemexpress LLC Supplier No. HY1310355MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lys-CoA TFA is a bisubstrate-mimic inhibitor of the p300 histone acetyltransferase (HAT). It inhibits p300-dependent acetylation (reported IC50 = 50-500 nM) with >100-fold selectivity over PCAF and is intended for use in biochemical and cell-based studies probing p300-mediated acetylation.

  • Selective p300 histone acetyltransferase (HAT) inhibitor (IC50 = 50-500 nM)
  • Greater than 100-fold selectivity versus PCAF
  • High purity (98.5%) suitable for research applications
  • Molecular formula C33H54F3N10O21P3S
  • Molecular weight 1108.82 g·mol⁻1
  • Supplied as a small-quantity solid for biochemical assays and cell studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.